Assay Details
| Gene Symbol: Ddit4l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080399.1 | NP_536324.1 | 224 | 95 | ||
| GenBank mRNA | AF327511.1 | - | 224 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF327511.1, NM_080399.1 transcripts.
Set Membership: |
|

Ddit4l2
Genomic Map