Assay Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246051.2 | XP_006246113.1 | 2 - 3 | 985 | 69 | |
| XM_006246052.2 | XP_006246114.1 | 2 - 3 | 649 | 69 | ||
| XM_006246053.2 | XP_006246115.1 | 2 - 3 | 435 | 69 | ||
| XM_008767606.1 | XP_008765828.1 | 2 - 3 | 985 | 69 | ||
More Information
Set Membership: |
|

Cramp1l
Genomic Map