Assay Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246051.2 | XP_006246113.1 | 4 - 5 | 1327 | 86 | |
| XM_006246052.2 | XP_006246114.1 | 4 - 5 | 991 | 86 | ||
| XM_006246053.2 | XP_006246115.1 | 4 - 5 | 777 | 86 | ||
| XM_008767606.1 | XP_008765828.1 | 4 - 5 | 1327 | 86 | ||
More Information
Set Membership: |
|

Cramp1l
Genomic Map