Assay Details
| Gene Symbol: Tril | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034010.1 | NP_001029182.1 | 1 | 395 | 53 | |
| GenBank mRNA | BC100659.1 | - | 1 | 395 | 7935751 | 53 |
More Information
Set Membership: |
|

Tril
Genomic Map