Assay Details
| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762874.1 | XP_008761096.1 | 8 | 3027 | 106 | |
| XM_008762875.1 | XP_008761097.1 | 8 | 5354 | 106 | ||
More Information
Set Membership: |
|

Wipf3
Genomic Map