Assay Details
| Gene Symbol: Qki | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758779.1 | XP_008757001.1 | 5923 | 96 | ||
| XM_008758780.1 | XP_008757002.1 | 4626 | 96 | |||
| XM_008758781.1 | XP_008757003.1 | 5899 | 96 | |||
More Information
Set Membership: |
|

Qki
Genomic Map