Assay Details
| Gene Symbol: Emp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012843.2 | NP_036975.1 | 2334 | 94 | ||
| XM_006237515.2 | XP_006237577.1 | 2443 | 94 | |||
| XM_008763369.1 | XP_008761591.1 | 2443 | 94 | |||
| GenBank mRNA | BC087031.1 | - | 2334 | 7192247 | 94 | |
More Information
Set Membership: |
|

Emp1
Genomic Map