Assay Details
| Gene Symbol: NEWGENE_1359502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237686.2 | XP_006237748.1 | 1 | 309 | 66 | |
| XM_006237687.2 | XP_006237749.1 | 1 | 309 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006237686.2, XM_006237687.2 transcripts.
Set Membership: |
|

NEWGENE_1359502
Genomic Map