Assay Details
| Gene Symbol: Slc35e2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239572.2 | XP_006239634.1 | 9 | 5619 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35e2b| Gene Symbol: Slc35e2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239572.2 | XP_006239634.1 | 9 | 5619 | 83 | |
Set Membership: |
|