Assay Details
| Gene Symbol: Hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173305.1 | NP_775427.1 | 2 - 3 | 578 | 87 | |
| XM_006240739.2 | XP_006240801.2 | 2 - 3 | 374 | 87 | ||
| GenBank mRNA | BC088104.1 | - | 2 - 3 | 528 | 7326228 | 87 |
| BC091114.1 | - | 2 - 3 | 490 | 7321553 | 87 | |
| U89280.1 | - | 2 - 3 | 578 | 87 | ||
| Gene Symbol: LOC100362350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769893.1 | XP_008768115.1 | 1 - 2 | 360 | 87 | |
More Information
Set Membership: |
|

Hsd17b6
Genomic Map