Assay Details
| Gene Symbol: Hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173305.1 | NP_775427.1 | 3 - 4 | 835 | 86 | |
| XM_006240739.2 | XP_006240801.2 | 3 - 4 | 631 | 86 | ||
| GenBank mRNA | BC088104.1 | - | 3 - 4 | 785 | 7326228 | 86 |
| BC091114.1 | - | 3 - 4 | 747 | 7321553 | 86 | |
| U89280.1 | - | 3 - 4 | 835 | 86 | ||
| Gene Symbol: LOC100362350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769893.1 | XP_008768115.1 | 2 - 3 | 617 | 86 | |
More Information
Set Membership: |
|

Hsd17b6
Genomic Map