Assay Details
| Gene Symbol: Hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173305.1 | NP_775427.1 | 4 - 5 | 998 | 84 | |
| XM_006240739.2 | XP_006240801.2 | 4 - 5 | 794 | 84 | ||
| GenBank mRNA | BC088104.1 | - | 4 - 5 | 948 | 7326228 | 84 |
| BC091114.1 | - | 4 - 5 | 910 | 7321553 | 84 | |
| U89280.1 | - | 4 - 5 | 998 | 84 | ||
| Gene Symbol: LOC100362350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769893.1 | XP_008768115.1 | 3 - 4 | 780 | 84 | |
More Information
Set Membership: |
|

Hsd17b6
Genomic Map