Assay Details
| Gene Symbol: Tmem200b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001068178.5 | XP_001068178.2 | 3 | 2231 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem200b| Gene Symbol: Tmem200b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001068178.5 | XP_001068178.2 | 3 | 2231 | 57 | |
Set Membership: |
|