Assay Details
| Gene Symbol: Ccdc71l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750141.3 | XP_003750189.2 | 1 | 680 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003750141.3 transcripts.
Set Membership: |
|

Ccdc71l
Genomic Map