Assay Details
| Gene Symbol: Sstr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012719.2 | NP_036851.1 | 2 | 1158 | 83 | |
| XM_006240115.2 | XP_006240177.1 | 2 | 2461 | 83 | ||
| GenBank mRNA | L02915.1 | - | 2 | 932 | 83 | |
| M97656.1 | - | 1 | 816 | 83 | ||
| X62314.1 | - | 1 | 840 | 83 | ||
More Information
Set Membership: |
|

Sstr1
Genomic Map