Assay Details
| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271791.1 | NP_001258720.1 | 1 | 3885 | 81 | |
| NR_073439.1 | 2 | 2855 | 81 | |||
| NR_073440.1 | 3 | 2644 | 81 | |||
| XM_008759646.1 | XP_008757868.1 | 2 | 4039 | 81 | ||
More Information
Set Membership: |
|

Crebzf
Genomic Map