Assay Details
| Gene Symbol: Zfp266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135018.1 | NP_001128490.1 | 9 | 4542 | 104 | |
| XM_008765961.1 | XP_008764183.1 | 8 | 5886 | 104 | ||
| XM_008765962.1 | XP_008764184.1 | 8 | 5894 | 104 | ||
| XM_008765963.1 | XP_008764185.1 | 7 | 5618 | 104 | ||
More Information
Set Membership: |
|

Zfp266
Genomic Map