Assay Details
| Gene Symbol: Chrnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U42976.1 | - | 2038 | 100 | ||
| X15834.1 | - | 2084 | 100 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Chrnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U42976.1 | - | 2038 | 100 | ||
| X15834.1 | - | 2084 | 100 | |||
Set Membership: |
|