Assay Details
| Gene Symbol: Blvrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106236.1 | NP_001099706.1 | 5 | 807 | 74 | |
| XM_008759117.1 | XP_008757339.1 | 5 | 728 | 74 | ||
| GenBank mRNA | BC168943.1 | - | 5 | 711 | 7322093 | 74 |
| FQ228106.1 | - | 5 | 691 | 74 | ||
| FQ230737.1 | - | 5 | 754 | 74 | ||
| FQ232598.1 | - | 5 | 754 | 74 | ||
More Information
Set Membership: |
|

Blvrb
Genomic Map