Assay Details
| Gene Symbol: Magea11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013962.1 | NP_001013984.1 | 3 | 1443 | 139 | |
| GenBank mRNA | BC083676.1 | - | 3 | 1443 | 7135789 | 139 |
More Information
Set Membership: |
|

Magea11
Genomic Map