Assay Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773695.1 | XP_008771917.1 | 21 - 22 | 2788 | 125 | |
| XM_008773696.1 | XP_008771918.1 | 19 - 20 | 2677 | 125 | ||
| GenBank mRNA | FJ775728.1 | - | 2578 | 125 | ||
| Gene Symbol: LOC103694558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773697.1 | XP_008771919.1 | 1804 | 125 | ||
| XM_008773698.1 | XP_008771920.1 | 1804 | 125 | |||
| XM_008773699.1 | XP_008771921.1 | 1669 | 125 | |||
| XM_008773700.1 | XP_008771922.1 | 1669 | 125 | |||
More Information
Set Membership: |
|

Kdm6a
Genomic Map