Assay Details
| Gene Symbol: LOC103694558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773697.1 | XP_008771919.1 | 2098 | 120 | ||
| XM_008773698.1 | XP_008771920.1 | 2098 | 120 | |||
| XM_008773699.1 | XP_008771921.1 | 1963 | 120 | |||
| XM_008773700.1 | XP_008771922.1 | 1963 | 120 | |||
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773695.1 | XP_008771917.1 | 24 - 25 | 3082 | 120 | |
| XM_008773696.1 | XP_008771918.1 | 22 - 23 | 2971 | 120 | ||
| GenBank mRNA | FJ775728.1 | - | 2872 | 120 | ||
More Information
Set Membership: |
|

LOC103694558
Genomic Map