Assay Details
| Gene Symbol: Tmem235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247851.2 | XP_006247913.1 | 2 - 3 | 1548 | 95 | |
| XM_006247852.2 | XP_006247914.1 | 2 - 3 | 488 | 95 | ||
| XM_008768444.1 | XP_008766666.1 | 1 - 2 | 1686 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247851.2, XM_006247852.2, XM_008768444.1 transcripts.
Set Membership: |
|

Tmem235
Genomic Map