Assay Details
| Gene Symbol: Tmem235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247851.2 | XP_006247913.1 | 4 - 5 | 2707 | 69 | |
| XM_006247852.2 | XP_006247914.1 | 4 - 5 | 1647 | 69 | ||
| XM_008768444.1 | XP_008766666.1 | 3 - 4 | 2845 | 69 | ||
More Information
Set Membership: |
|

Tmem235
Genomic Map