Assay Details
| Gene Symbol: Linc00514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767626.1 | XP_008765848.1 | 17 - 18 | 656 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Linc00514| Gene Symbol: Linc00514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767626.1 | XP_008765848.1 | 17 - 18 | 656 | 66 | |
Set Membership: |
|