Assay Details
| Gene Symbol: Krtap3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001055567.3 | XP_001055567.1 | 1 | 218 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap3-1| Gene Symbol: Krtap3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001055567.3 | XP_001055567.1 | 1 | 218 | 99 | |
Set Membership: |
|