Assay Details
| Gene Symbol: LOC100363287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002727853.3 | XP_002727899.3 | 1 | 119 | 61 | |
| Gene Symbol: Krtap13-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109325.1 | NP_001102795.1 | 1 | 155 | 61 | |
| Gene Symbol: Krtap14l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248116.2 | XP_006248178.1 | 1 | 101 | 61 | |
| Gene Symbol: LOC100363236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751020.3 | XP_003751068.2 | 1 | 101 | 61 | |
More Information
Set Membership: |
|

LOC100363287
Genomic Map