Assay Details
| Gene Symbol: Pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025272.1 | NP_001020443.1 | 1 - 2 | 223 | 71 | |
| XM_006249534.2 | XP_006249596.1 | 1 - 2 | 255 | 71 | ||
| GenBank mRNA | BC092607.1 | - | 1 - 2 | 223 | 7309810 | 71 |
More Information
Set Membership: |
|

Pgam5
Genomic Map