Assay Details
| Gene Symbol: Rnf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113748.1 | NP_001107220.1 | 1 | 1984 | 84 | |
| GenBank mRNA | BC158712.1 | - | 1 | 1984 | 9035415 | 84 |
More Information
Set Membership: |
|

Rnf26
Genomic Map