Assay Details
| Gene Symbol: RGD1564236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245569.2 | XP_006245631.1 | 1 | 623 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006245569.2 transcripts.
Set Membership: |
|

RGD1564236
Genomic Map