Assay Details
| Gene Symbol: LOC102549694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245564.2 | XP_006245626.1 | 1 | 2430 | 73 | |
| XM_006245565.2 | XP_006245627.1 | 1 | 2430 | 73 | ||
| XM_006245566.2 | XP_006245628.1 | 1 | 2430 | 73 | ||
| XM_006245567.2 | XP_006245629.1 | 1 | 2430 | 73 | ||
More Information
Set Membership: |
|

LOC102549694
Genomic Map