Assay Details
| Gene Symbol: Mafg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022386.2 | NP_071781.1 | 1 - 2 | 162 | 103 | |
| GenBank mRNA | BC078828.1 | - | 1 - 2 | 162 | 7127046 | 103 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078828.1, NM_022386.2 transcripts.
Set Membership: |
|

Mafg
Genomic Map