Assay Details
| Gene Symbol: Mnda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012029.1 | NP_001012029.1 | 2 - 3 | 222 | 101 | |
| GenBank mRNA | BC085891.1 | - | 2 - 3 | 222 | 7192620 | 101 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085891.1, NM_001012029.1 transcripts.
Set Membership: |
|

Mnda
Genomic Map