Assay Details
| Gene Symbol: Mnda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012029.1 | NP_001012029.1 | 6 - 7 | 1261 | 120 | |
| GenBank mRNA | BC085891.1 | - | 6 - 7 | 1261 | 7192620 | 120 |
More Information
Set Membership: |
|

Mnda
Genomic Map