Assay Details
| Gene Symbol: Fam71e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145401.1 | NP_001138873.1 | 2 - 3 | 473 | 64 | |
| XM_008758921.1 | XP_008757143.1 | 2 - 3 | 818 | 64 | ||
| XM_008758922.1 | XP_008757144.1 | 2 - 3 | 818 | 64 | ||
| GenBank mRNA | BC161877.1 | - | 2 - 3 | 473 | 64 | |
More Information
Set Membership: |
|

Fam71e2
Genomic Map