Assay Details
| Gene Symbol: Cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012830.1 | NP_036962.1 | 1 - 2 | 147 | 72 | |
| GenBank mRNA | BC088164.1 | - | 1 - 2 | 77 | 7374969 | 72 |
| X05111.1 | - | 1 - 2 | 147 | 72 | ||
More Information
Set Membership: |
|

Cd2
Genomic Map