Assay Details
| Gene Symbol: Cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012830.1 | NP_036962.1 | 2 - 3 | 452 | 89 | |
| GenBank mRNA | BC088164.1 | - | 2 - 3 | 382 | 7374969 | 89 |
| X05111.1 | - | 2 - 3 | 452 | 89 | ||
More Information
Set Membership: |
|

Cd2
Genomic Map