Assay Details
| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014198.2 | NP_001014220.2 | 1 - 2 | 85 | 92 | |
| GenBank mRNA | BC087681.1 | - | 1 - 2 | 62 | 7321205 | 92 |
| FM111441.1 | - | 1 - 2 | 88 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC087681.1, NM_001014198.2 transcripts.
Set Membership: |
|

Gskip
Genomic Map