Assay Details
| Gene Symbol: Padi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017226.1 | NP_058922.1 | 15 - 16 | 1825 | 61 | |
| XM_008764215.1 | XP_008762437.1 | 16 - 17 | 1929 | 61 | ||
| GenBank mRNA | J05022.1 | - | 15 - 16 | 1825 | 61 | |
More Information
Set Membership: |
|

Padi2
Genomic Map