Assay Details
| Gene Symbol: Gtf2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022208.1 | NP_071544.1 | 6 - 7 | 616 | 60 | |
| XM_006240410.2 | XP_006240472.1 | 6 - 7 | 891 | 60 | ||
| GenBank mRNA | AF000943.1 | - | 6 - 7 | 616 | 60 | |
More Information
Set Membership: |
|

Gtf2a1
Genomic Map