Assay Details
| Gene Symbol: Fgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022211.1 | NP_071547.1 | 1 - 2 | 346 | 76 | |
| GenBank mRNA | D64085.1 | - | 1 - 2 | 346 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Fgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022211.1 | NP_071547.1 | 1 - 2 | 346 | 76 | |
| GenBank mRNA | D64085.1 | - | 1 - 2 | 346 | 76 | |
Set Membership: |
|