Assay Details
| Gene Symbol: Rhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022505.2 | NP_071950.1 | 2 - 3 | 365 | 69 | |
| XM_006239268.2 | XP_006239330.1 | 2 - 3 | 399 | 69 | ||
| GenBank mRNA | AB015191.1 | - | 2 - 3 | 330 | 69 | |
| AF531096.1 | - | 2 - 3 | 365 | 69 | ||
| BC127471.1 | - | 2 - 3 | 354 | 8374048 | 69 | |
More Information
Set Membership: |
|

Rhd
Genomic Map