Assay Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030867.2 | NP_110494.2 | 3 - 4 | 881 | 86 | |
| GenBank mRNA | AF246634.1 | - | 3 - 4 | 704 | 86 | |
| BC085729.1 | - | 3 - 4 | 881 | 7114624 | 86 | |
| FQ235103.1 | - | 3 - 4 | 827 | 86 | ||
More Information
Set Membership: |
|

Nfkbib
Genomic Map