Assay Details
| Gene Symbol: Wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031590.1 | NP_113778.1 | 3 - 4 | 540 | 97 | |
| GenBank mRNA | AF259981.1 | - | 3 - 4 | 540 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031590.1 | NP_113778.1 | 3 - 4 | 540 | 97 | |
| GenBank mRNA | AF259981.1 | - | 3 - 4 | 540 | 97 | |
Set Membership: |
|