Assay Details
| Gene Symbol: Wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031590.1 | NP_113778.1 | 4 - 5 | 794 | 88 | |
| GenBank mRNA | AF259981.1 | - | 4 - 5 | 794 | 88 | |
| BC089187.1 | - | 3 - 4 | 88 | |||
More Information
Set Membership: |
|

Wisp2
Genomic Map