Assay Details
| Gene Symbol: Hsd17b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031682.1 | NP_113870.1 | 5 - 6 | 607 | 73 | |
| GenBank mRNA | AF049878.1 | - | 5 - 6 | 595 | 73 | |
| AF069770.1 | - | 5 - 6 | 607 | 73 | ||
| BC158587.1 | - | 5 - 6 | 601 | 9089087 | 73 | |
| FQ210583.1 | - | 3 - 4 | 348 | 73 | ||
| FQ220319.1 | - | 3 - 4 | 348 | 73 | ||
| FQ225948.1 | - | 3 - 4 | 348 | 73 | ||
| FQ228203.1 | - | 3 - 4 | 348 | 73 | ||
| FQ230672.1 | - | 3 - 4 | 349 | 73 | ||
| FQ231826.1 | - | 3 - 4 | 348 | 73 | ||
| FQ232016.1 | - | 3 - 4 | 348 | 73 | ||
More Information
Set Membership: |
|

Hsd17b10
Genomic Map