Assay Details
| Gene Symbol: Rab9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053458.1 | NP_445910.1 | 2 - 3 | 147 | 97 | |
| GenBank mRNA | AF325692.1 | - | 2 - 3 | 147 | 97 | |
| BC070502.1 | - | 2 - 3 | 151 | 7096737 | 97 | |
| FQ212962.1 | - | 2 - 3 | 156 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF325692.1, BC070502.1, NM_053458.1 transcripts.
Set Membership: |
|

Rab9a
Genomic Map