Assay Details
| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057199.1 | NP_476547.1 | 7 - 8 | 1035 | 95 | |
| GenBank mRNA | AF082533.1 | - | 7 - 8 | 1035 | 95 | |
| AJ012741.1 | - | 7 - 8 | 1025 | 95 | ||
More Information
Set Membership: |
|

Ncr1
Genomic Map