Assay Details
| Gene Symbol: Rhov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138542.2 | NP_612551.2 | 1 - 2 | 31 | 74 | |
| GenBank mRNA | BC086990.1 | - | 1 - 2 | 31 | 7135643 | 74 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086990.1, NM_138542.2 transcripts.
Set Membership: |
|

Rhov
Genomic Map