Assay Details
| Gene Symbol: Kir3dl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181479.2 | NP_852144.1 | 6 - 7 | 1080 | 129 | |
| GenBank mRNA | AF527797.1 | - | 6 - 7 | 1080 | 129 | |
More Information
Set Membership: |
|

Kir3dl1
Genomic Map